Thermo Scientific Chemicals

Copper(I) cyanide, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 202080010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(I) cyanide, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 202080010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202080010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
544-92-3
IUPAC Name
λ¹-copper(1+) iminomethanide
Molecular Formula
CCuN
InChI Key
DOBRDRYODQBAMW-UHFFFAOYSA-N
SMILES
[Cu+].[C-]#N
Arsenic (As)
=<10 ppm
Appearance (Color)
White to beige to green
Appearance (Form)
Powder
Assay
>=98.5 % (ex Cu)
Iron (Fe)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text