Thermo Scientific Chemicals

Zinc bromide, 98%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 202100025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Zinc bromide, 98%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 202100025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 202100025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7699-45-8
IUPAC Name
zinc(2+) dibromide
Molecular Formula
Br2Zn
InChI Key
VNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES
[Zn++].[Br-].[Br-]
Appearance (Color)
White to off-white
Barium (Ba)
=<10 ppm
Iron (Fe)
=<10 ppm
Potassium (K)
=<100 ppm
Appearance (Form)
Powder and/or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text