Thermo Scientific™

Pierce™ GST Agarose

Catalog number: 20211
Have Questions?
Thermo Scientific™

Pierce™ GST Agarose

Catalog number: 20211
Have Questions?
Catalog Number
20211
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
202115 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Immobilized GST is glutathione S-transferase from Schistosoma japonicum that has been covalently immobilized on crosslinked, 6% beaded agarose.

Features of GST Agarose:

• Provides purification of specific antibodies from those directed against the GST tag
• Reusable—bound antibodies can be eluted with 6 M guanidinium chloride allowing for 10 purifications with the support

Antibody molecules recognizing the GST portion of a recombinant fusion protein can be removed from the desired antibody by passing antiserum over an immobilized GST column. GST antibodies will bind to the immobilized GST column, whereas antibodies against other fusion proteins will pass through the column unrestrained. The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins.

Related Products
Pierce™ GST Agarose Columns, 2 mL
Pierce™ GST (glutathione-S-transferase)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Agarose
Product Line
Pierce™
Column Type
Agarose Resin, Affinity
Format
Bottle
Stationary Phase
GST Protein
Purification Target
Anti-GST Antibody
Description
Pierce GST Agarose
Quantity
5 mL

Contents & Storage

4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text