Thermo Scientific Chemicals

Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 202110250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 202110250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202110250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9042-11-9
MDL Number
MFCD00132693
Appearance (Form)
Powder
Particle size
=<10 % (< 75 µm)
Appearance (Color)
White to off-white
Total weight capacity
>=10 meq/g
Particle size
=<10 % (> 150 µm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text