Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals
Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number 202111000
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific Chemicals
Catalog number202111000
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS9042-11-9
MDL NumberMFCD00132693
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Particle size=<10 % (< 75 µm)
Appearance (Color)White to off-white
Total weight capacity>=10 meq/g
Particle size=<10 % (> 150 µm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only