Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals
Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 202172500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9049-93-8
MDL NumberMFCD00145842
SpecificationsSpecification SheetSpecification Sheet
Particle size=<2.0 % (> 1.180 mm)
Appearance (Color)Light brown
Harmonic mean size0.490 to 0.690 mm
Particle size=<1.0 % (< 0.300 mm)
Total volume capacity>=1.3 eq/l (free base)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only