Amberlyst™ A26, OH-form, ion-exchange resin, Thermo Scientific Chemicals
Amberlyst™ A26, OH-form, ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlyst™ A26, OH-form, ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Plastic bottle
Catalog number 202181000
Price (USD)/ Each
50.65
Online exclusive
59.50 
Save 8.85 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
50.65
Online exclusive
59.50 
Save 8.85 (15%)
Add to cart
Amberlyst™ A26, OH-form, ion-exchange resin, Thermo Scientific Chemicals
Catalog number202181000
Price (USD)/ Each
50.65
Online exclusive
59.50 
Save 8.85 (15%)
-
Add to cart
Chemical Identifiers
MDL NumberMFCD00145579
CAS39339-85-0
SpecificationsSpecification SheetSpecification Sheet
Uniformity coefficient=<1.45
Harmonic mean size0.560 to 0.700
Appearance (Color)Tan
Total volume capacity>=0.80 eq/l (OH) as is
Appearance (Form)Beads
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only