Thermo Scientific Chemicals

Amberlite™ XAD-4, Thermo Scientific Chemicals

Catalog number: 202230025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Amberlite™ XAD-4, Thermo Scientific Chemicals

Catalog number: 202230025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202230025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9003-69-4
MDL Number
MFCD00132704
Moisture holding capacity
54.0 to 60.0 %
Uniformity coefficient
=<2.0
Particle size
=<5.0 % (< 0.300 mm)
Appearance (Color)
White to yellow
Appearance (Form)
Beads

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text