Thermo Scientific Chemicals

Amberlite™ XAD-7 HP, Thermo Scientific Chemicals

Catalog number: 202240025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Amberlite™ XAD-7 HP, Thermo Scientific Chemicals

Catalog number: 202240025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202240025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
37380-43-1
MDL Number
MFCD00132705
TOC-AFNOR test
=<10 ppm (total)
Appearance (Color)
White to pink-beige
Moisture holding capacity
61.0 to 69.0 %
Harmonic mean size
0.430 to 0.690 mm
Particle size
=<8.0 % (> 1.180 mm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text