Amberlite™ XAD-7 HP, Thermo Scientific Chemicals
Amberlite™ XAD-7 HP, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ XAD-7 HP, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 202245000
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
Add to cart
Amberlite™ XAD-7 HP, Thermo Scientific Chemicals
Catalog number202245000
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS37380-43-1
MDL NumberMFCD00132705
SpecificationsSpecification SheetSpecification Sheet
TOC-AFNOR test=<10 ppm (total)
Appearance (Color)White to pink-beige
Moisture holding capacity61.0 to 69.0 %
Harmonic mean size0.430 to 0.690 mm
Particle size=<8.0 % (> 1.180 mm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only