Ammonium hexafluorophosphate, 99%, pure, Thermo Scientific Chemicals
Ammonium hexafluorophosphate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hexafluorophosphate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 202320050
Price (USD)/ Each
20.65
Online exclusive
24.30 
Save 3.65 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
20.65
Online exclusive
24.30 
Save 3.65 (15%)
Add to cart
Ammonium hexafluorophosphate, 99%, pure, Thermo Scientific Chemicals
Catalog number202320050
Price (USD)/ Each
20.65
Online exclusive
24.30 
Save 3.65 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameammonium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6H4NP
InChI KeyNIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES[NH4+].F[P-](F)(F)(F)(F)F
Molecular Weight (g/mol)163.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Elemental Analysis>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only