Potassium chromate, 99.5%, for analysis
Potassium chromate, 99.5%, for analysis
Potassium chromate, 99.5%, for analysis
Potassium chromate, 99.5%, for analysis
Thermo Scientific Chemicals

Potassium chromate, 99.5%, for analysis

CAS: 7789-00-6 | CrK2O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202340025
Price (USD)
379.65
Special offer
Online exclusive
Ends: 26-Sep-2025
468.00
Save 88.35 (19%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
379.65
Special offer
Online exclusive
Ends: 26-Sep-2025
468.00
Save 88.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric>=99.5 %
Iron (Fe)=<20 ppm
Appearance (Form)Crystals
Sulfate (SO4)=<100 ppm
Calcium (Ca)=<50 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473780Certificate of AnalysisJun 25, 2025202340025, 202345000
A0471179Certificate of AnalysisMay 17, 2025202340025, 202345000
A0442740Certificate of AnalysisJan 31, 2025202340025, 202345000
A0464040Certificate of AnalysisJul 02, 2024202340025, 202345000
A0459025Certificate of AnalysisJan 03, 2024202340025, 202345000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection