Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Tetrakis(triphenylphosphine)palladium(0), 99%
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99%

CAS: 14221-01-3 | C72H60P4Pd | 1155.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 202380050
Price (USD)/ Each
126.65
Special offer
Online exclusive
149.00
Save 22.35 (15%)
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
126.65
Special offer
Online exclusive
149.00
Save 22.35 (15%)
Add to cart
Chemical Identifiers
IUPAC Nametetrakis(triphenylphosphane) palladium
Molecular FormulaC72H60P4Pd
InChI KeyNFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)1155.59
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Infrared spectrumConforms
Carbon (C)73.7 to 76.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link

1x1 image pixel for data collection