Silver acetate, 99%, pure, Thermo Scientific Chemicals
Silver acetate, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver acetate, 99%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 202390250
Price (USD)/ Each
153.65
Online exclusive
181.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
181.00 
Save 27.35 (15%)
Add to cart
Silver acetate, 99%, pure, Thermo Scientific Chemicals
Catalog number202390250
Price (USD)/ Each
153.65
Online exclusive
181.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesilver(1+) acetate
Molecular FormulaC2H3AgO2
InChI KeyCQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES[Ag+].CC([O-])=O
Molecular Weight (g/mol)166.91
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light grey
Appearance (Form)Powder
Titration Argentometric>=98.5 %
Infrared spectrumConforms

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only