Thermo Scientific Chemicals

Silver acetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 202391000
100 g, Each
Thermo Scientific Chemicals

Silver acetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 202391000
100 g, Each
Quantity
Catalog number: 202391000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-63-3
IUPAC Name
silver(1+) acetate
Molecular Formula
C2H3AgO2
InChI Key
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES
[Ag+].CC([O-])=O
Appearance (Color)
Off-white to beige to light grey
Appearance (Form)
Powder
Titration Argentometric
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text