Tannic acid, 95%, Thermo Scientific Chemicals
Tannic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tannic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
500 g
5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202420010
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
Add to cart
Tannic acid, 95%, Thermo Scientific Chemicals
Catalog number202420010
Price (USD)/ Each
141.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
167.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate
Molecular FormulaC76H52O46
InChI KeyLRBQNJMCXXYXIU-UHFFFAOYNA-N
SMILESOC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1
Molecular Weight (g/mol)1701.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
Loss on drying=<12.0 %
Appearance (Form)Powder
Assay>=94 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only