Thermo Scientific Chemicals

Calcon, pure

Catalog number: 202441000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Calcon, pure

Catalog number: 202441000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 202441000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2538-85-4
IUPAC Name
sodium (4E)-4-[2-(2-hydroxynaphthalen-1-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Molecular Formula
C20H13N2NaO5S
InChI Key
YJUGDMOGPJJWDG-QPNALZDCSA-M
SMILES
[Na+].OC1=CC=C2C=CC=CC2=C1N\N=C1\C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O
Appearance (Form)
Powder
Appearance (Color)
Dark brown to black
Infrared spectrum
Conforms
UV spectrum
Complies

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text