Thermo Scientific Chemicals

Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized, Thermo Scientific Chemicals

Catalog number: 202460010
1 L, Plastic bottle, Each
Thermo Scientific Chemicals

Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized, Thermo Scientific Chemicals

Catalog number: 202460010
1 L, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202460010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7722-84-1
IUPAC Name
peroxol
Molecular Formula
H2O2
InChI Key
MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES
OO
Appearance (Form)
Clear solution
Heavy metals
=<10 ppm
Chloride (Cl)
=<2 ppm
Stabilizer
about 0.03% stabilizers
Color scale
=<10 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text