Thermo Scientific Chemicals

6-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 202540250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: 202540250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 202540250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3167-49-5
IUPAC Name
6-aminopyridine-3-carboxylic acid
Molecular Formula
C6H6N2O2
InChI Key
ZCIFWRHIEBXBOY-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=N1)C(O)=O
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %
Appearance (Color)
White to beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text