Thermo Scientific Chemicals

8-Azahypoxanthine, 98%, Thermo Scientific Chemicals

Catalog number: 202590010
1 g, Each
Thermo Scientific Chemicals

8-Azahypoxanthine, 98%, Thermo Scientific Chemicals

Catalog number: 202590010
1 g, Each
Quantity
Catalog number: 202590010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2683-90-1
IUPAC Name
2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
Molecular Formula
C4H3N5O
InChI Key
OEEYCNOOAHGFHL-UHFFFAOYSA-N
SMILES
O=C1N=CN=C2NNN=C12
Appearance (Form)
Crystalline powder
HPLC
>=97.5 %
Appearance (Color)
Light yellow to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text