Thermo Scientific Chemicals

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%, Thermo Scientific Chemicals

Catalog number: 202640250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%, Thermo Scientific Chemicals

Catalog number: 202640250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 202640250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64431-96-5
IUPAC Name
2-[(3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C11H26N2O6
InChI Key
HHKZCCWKTZRCCL-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)NCCCNC(CO)(CO)CO
Infrared spectrum
Conforms
Titration with HClO4
98.5 to 101.5 %
UV
(0.1 M in water)
UV
at 400 nm A: =<0.05
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text