Thermo Scientific Chemicals

Hexamethyldisilane, 98+%, Thermo Scientific Chemicals

Catalog number: 202692500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Hexamethyldisilane, 98+%, Thermo Scientific Chemicals

Catalog number: 202692500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 202692500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1450-14-2
IUPAC Name
hexamethyldisilane
Molecular Formula
C6H18Si2
InChI Key
NEXSMEBSBIABKL-UHFFFAOYSA-N
SMILES
C[Si](C)(C)[Si](C)(C)C
Appearance (Form)
Liquid
GC
>=98.0 %
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4211 to 1.4231 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text