Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 202850100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 202850100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 202850100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10102-17-7
IUPAC Name
disodium pentahydrate sulfanidesulfonate
Molecular Formula
H10Na2O8S2
InChI Key
PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
SO4 and SO3 (as SO4)
=<0.1 %
Insoluble matter
=<0.005 % (in water)
Nitrogen compounds (as N)
=<0.002 %
Sulfur (S)
=<1 ppm
Chloride (Cl)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text