Thermo Scientific Chemicals

Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 202860051
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 202860051
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 202860051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6381-59-5
IUPAC Name
potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate
Molecular Formula
C4H12KNaO10
InChI Key
VZOPRCCTKLAGPN-ZFJVMAEJSA-L
SMILES
O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Infrared spectrum
Conforms
Iron (Fe)
=<10 ppm
Appearance (Color)
Semitransparent white
Titration with HClO4
99.0 to 102.0 %
Chloride (Cl)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text