Trichloroacetyl isocyanate, NMR grade, Thermo Scientific Chemicals
Trichloroacetyl isocyanate, NMR grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trichloroacetyl isocyanate, NMR grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 202900020
Price (USD)/ Each
46.90
-
Add to cart
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
46.90
Add to cart
Trichloroacetyl isocyanate, NMR grade, Thermo Scientific Chemicals
Catalog number202900020
Price (USD)/ Each
46.90
-
Add to cart
Chemical Identifiers
CAS3019-71-4
IUPAC Nametrichloroethanecarbonyl isocyanate
Molecular FormulaC3Cl3NO2
InChI KeyGRNOZCCBOFGDCL-UHFFFAOYSA-N
SMILESClC(Cl)(Cl)C(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4799 to 1.4819 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only