2,2,2-Trichloroacetamide, 98+%, Thermo Scientific Chemicals
2,2,2-Trichloroacetamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trichloroacetamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 202921000
Price (USD)/ Each
68.50
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
68.50
Add to cart
2,2,2-Trichloroacetamide, 98+%, Thermo Scientific Chemicals
Catalog number202921000
Price (USD)/ Each
68.50
-
Add to cart
Chemical Identifiers
CAS594-65-0
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.0 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Powder or crystals and/or chunks
Melting point139°C to 143°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only