Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Thermo Scientific Chemicals

Dowex™ 1X2, 100-200 mesh, ion-exchange resin

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
Have Questions?
Catalog NumberQuantity
2029600252.5 kg
Catalog number 202960025
Price (USD)
1,118.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
1,118.00
Each
Add to cart
Chemical Identifiers
CAS9085-42-1
MDL NumberMFCD00132715
SpecificationsSpecification SheetSpecification Sheet
Water retention capacity65 to 75 % (Cl)
Particle size=<5 % (< 100 µm)
Appearance (Form)Beads
Particle size100 to 200 mesh
Particle size=<10 % (> 300 µm)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0410048Certificate of AnalysisJan 04, 2024202961000, 202960025, 202965000
A0320991Certificate of AnalysisDec 13, 2022202961000, 202960025, 202965000
A0384827Certificate of AnalysisMay 24, 2021202961000, 202960025, 202965000
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection