Thermo Scientific Chemicals

Dowex™ 1X2-400, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 202971000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Dowex™ 1X2-400, ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 202971000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202971000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9085-42-1
MDL Number
MFCD00132715
Appearance (Form)
Beads
Water retention capacity
65 to 75 % (Cl)
Particle size
200 to 400 mesh
Particle size
=<15 % (> 210 µm)
Wet volume capacity
>=0.6 meq/ml (Cl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text