Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 203130010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 203130010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 203130010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10326-27-9
IUPAC Name
barium(2+) dihydrate dichloride
Molecular Formula
BaCl2H4O2
InChI Key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ba++]
Titration Argentometric
>=99.0 %
Loss on drying
14.0 % to 16.0 % (150°C)
pH
5.2 to 8.2 (5 % at 25°C)
Heavy metals (as Pb)
=<5 ppm
Calcium (Ca)
=<500 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text