Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 203130025
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
Add to cart
Barium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number203130025
Price (USD)/ Each
178.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
210.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namebarium(2+) dihydrate dichloride
Molecular FormulaBaCl2H4O2
InChI KeyPWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ba++]
Molecular Weight (g/mol)244.26
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Loss on drying14.0 % to 16.0 % (150°C)
pH5.2 to 8.2 (5 % at 25°C)
Heavy metals (as Pb)=<5 ppm
Calcium (Ca)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only