Barium hydroxide octahydrate, 98+%, for analysis, Thermo Scientific Chemicals
Barium hydroxide octahydrate, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium hydroxide octahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Barium hydroxide octahydrate, 98+%, for analysis, ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, CAS: 12230-71-6, White, Grade: Analysis, Melting Point: 78.0 deg.C, Molecular Formula: H2BaO2.8H2O, Molecular Weight: 315.48, Percent Purity: 98+%
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 203142500
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
Add to cart
Barium hydroxide octahydrate, 98+%, for analysis, Thermo Scientific Chemicals
Catalog number203142500
Price (USD)/ Each
32.65
Online exclusive
36.40 
Save 3.75 (10%)
-
Add to cart
Chemical Identifiers
CAS12230-71-6
IUPAC Namebarium(2+) octahydrate dihydroxide
Molecular FormulaBaH18O10
InChI KeyZUDYPQRUOYEARG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or flakes
Appearance (Color)White
Titration with HCl>=98.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.01 % (in HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only