Thermo Scientific Chemicals

Adipic dihydrazide, 98%, Thermo Scientific Chemicals

Catalog number: 203310250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Adipic dihydrazide, 98%, Thermo Scientific Chemicals

Catalog number: 203310250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 203310250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1071-93-8
IUPAC Name
hexanedihydrazide
Molecular Formula
C6H14N4O2
InChI Key
IBVAQQYNSHJXBV-UHFFFAOYSA-N
SMILES
NNC(=O)CCCCC(=O)NN
Appearance (Color)
White to light yellow
Titration
>=97.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
178°C to 182°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text