Thermo Scientific Chemicals

Copper(II) fluoride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog number: 203371000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) fluoride, 98%, anhydrous, Thermo Scientific Chemicals

Catalog number: 203371000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 203371000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-19-7
IUPAC Name
copper(2+) difluoride
Molecular Formula
CuF2
InChI Key
GWFAVIIMQDUCRA-UHFFFAOYSA-L
SMILES
[F-].[F-].[Cu++]
Assay
>=97.5 %
Appearance (Color)
Light grey to blue-grey
Appearance (Form)
Powder
Loss on drying
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text