Vincristine sulfate, 95%
Vincristine sulfate, 95%
Thermo Scientific Chemicals

Vincristine sulfate, 95%

 
Have Questions?
Quantity:
5 mg
Packaging:
Glass Bottle
Catalog number 203440050
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
194.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
5 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
194.00 
Save 29.35 (15%)
Add to cart
Vincristine sulfate, 95%
Catalog number203440050
Price (USD)/ Each
164.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
194.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
Molecular FormulaC46H58N4O14S
InChI KeyAQTQHPDCURKLKT-JKDPCDLQSA-N
SMILESOS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C=O)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
Molecular Weight (g/mol)923.04
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<12 %
Appearance (Color)White to light yellow
HPLC>=94 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only