Thermo Scientific Chemicals

Vinblastine sulfate, 96.0-102.0%

Catalog number: 203450250
25 mg, Glass bottle, Each
Thermo Scientific Chemicals

Vinblastine sulfate, 96.0-102.0%

Catalog number: 203450250
25 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 203450250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
143-67-9
IUPAC Name
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
Molecular Formula
C46H60N4O13S
InChI Key
KDQAABAKXDWYSZ-JKDPCDLQSA-N
SMILES
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
Appearance (Color)
White to light yellow
Water
=<15 %
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
HPLC
96.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text