Informational:New Protein Purification and Isolation Support Center—Find technical tips and troubleshooting recommendations from our scientists.
Thermo Scientific™

Pierce™ Streptavidin Agarose

Catalog number: 20347
Have Questions?
Thermo Scientific™

Pierce™ Streptavidin Agarose

Catalog number: 20347
Have Questions?
Catalog Number
20347
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
203472 mL
-
2035310 mL
-
203495 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Streptavidin Agarose is a standard-capacity beaded agarose resin of immobilized recombinant streptavidin protein for use in a variety of biotin-binding affinity purification methods.

Features of Streptavidin Agarose:

Streptavidin—purified recombinant streptavidin protein (53kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
Agarose resin—support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable—streptavidin is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
Standard capacity—this variety of beads has a normal load of immobilized streptavidin, providing a binding capacity of 1 to 3 mg of biotinylated BSA per mL of resin

Properties of crosslinked 6% beaded agarose (CL-6B):
• Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
• Average Particle Size: 45 to 165 microns
• Exclusion Limit: 10,000 to 4,000,000 daltons
• Maximum Volumetric Flow Rate: approx. 1 mL/minute (for 1 cm diameter column)
• Maximum Linear Velocity: 30 cm per hour
• Maximum Pressure: less than 25psi (1.5 bar)

Applications:

• Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies
• Isolation of cell-surface proteins that have been labeled with a cleavable biotinylation reagent
• Isolation of cell-surface glycoproteins using biotinylated Concanavalin A
• Recovery of single-stranded biotinylated DNA for dideoxy sequencing
• Streptavidin-coated plates used for direct solid-phase sequencing
In vitro mutagenesis with streptavidin-coated plates
• DNA fingerprinting with streptavidin-coated plates

Pierce Streptavidin Agarose consists of purified recombinant streptavidin that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. Streptavidin is a tetrameric biotin-binding protein. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity procedures. Biotinylated antibodies, proteins, peptides, nucleic acids and other molecules or interaction complexes can be captured, immunoprecipitated, removed or purified from samples using this streptavidin resin.

Streptavidin is a tetrameric protein containing four biotin-binding sites similar to avidin. The native protein from Streptomyces avidinii is carbohydrate-free, has an acidic pI of 5.5 and a mass of 75 kDa. The recombinant protein used to manufacture Thermo Scientific Pierce Streptavidin Resins has a near-neutral isoelectric point and a mass of 53kDa. Streptavidin generally has less nonspecific binding than avidin (from chicken egg white) because of the absence of carbohydrates.

Related Products
Pierce™ Streptavidin Agarose Columns, 1 mL
Pierce™ High Capacity Streptavidin Agarose
Pierce™ Streptavidin Plus UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
1 to 3 mg Biotinylated BSA Binding per mL Resin
Particle Size
45 to 165 μm
Product Line
Pierce™
Quantity
2 mL
Protein Form
Recombinant
Target
Biotin-Tagged Molecule
Type
Agarose
Stationary Phase
Streptavidin
Sufficient For
Binding 15 to 28 μg Biotin/mL Resin (1 to 3 mg Biotinylated BSA/mL Resin)
Format
Bottle
Column Type
Affinity, Agarose Resin

Contents & Storage

Upon receipt store at 4°C. Do not freeze the resin. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text