DL-3-Octanol, 97%
DL-3-Octanol, 97%
DL-3-Octanol, 97%
Thermo Scientific Chemicals

DL-3-Octanol, 97%

CAS: 589-98-0 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 203770500
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
50.80
Save 9.15 (18%)
Each
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 26-Sep-2025
50.80
Save 9.15 (18%)
Each
Add to cart
Chemical Identifiers
CAS589-98-0
IUPAC Nameoctan-3-ol
Molecular FormulaC8H18O
InChI KeyNMRPBPVERJPACX-UHFFFAOYNA-N
SMILESCCCCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4230 to 1.4320 (20°C, 589 nm)
Density(20°C) 0.817 to 0.827 kg/l
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link