Thermo Scientific Chemicals

Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals

Catalog number: 203821000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals

Catalog number: 203821000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 203821000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
681-84-5
IUPAC Name
tetramethyl silicate
Molecular Formula
C4H12O4Si
InChI Key
LFQCEHFDDXELDD-UHFFFAOYSA-N
SMILES
CO[Si](OC)(OC)OC
Color scale
=<20 APHA
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms
Refractive index
1.3670 to 1.3700 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text