Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals
Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 203825000
Price (USD)/ Each
212.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
250.00 
Save 37.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
212.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
250.00 
Save 37.35 (15%)
Add to cart
Tetramethyl orthosilicate, 99%, Thermo Scientific Chemicals
Catalog number203825000
Price (USD)/ Each
212.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
250.00 
Save 37.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametetramethyl silicate
Molecular FormulaC4H12O4Si
InChI KeyLFQCEHFDDXELDD-UHFFFAOYSA-N
SMILESCO[Si](OC)(OC)OC
Molecular Weight (g/mol)152.22
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.3670 to 1.3700 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only