Thermo Scientific Chemicals

Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals

Catalog number: 203925000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium hexacyanocobaltate(III), 95%, Thermo Scientific Chemicals

Catalog number: 203925000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 203925000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13963-58-1
IUPAC Name
cobalt(3+) tripotassium hexakis(iminomethanide)
Molecular Formula
C6CoK3N6
InChI Key
LGRDAQPMSDIUQJ-UHFFFAOYSA-N
SMILES
[K+].[K+].[K+].[Co+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Assay
>=94 %
Appearance (Form)
Crystalline solid
Appearance (Color)
Light yellow to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text