Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 204030500
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
Add to cart
Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals
Catalog number204030500
Price (USD)/ Each
264.65
Online exclusive
294.00 
Save 29.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2001-45-8
IUPAC Nametetraphenylphosphanium chloride
Molecular FormulaC24H20ClP
InChI KeyWAGFXJQAIZNSEQ-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Titration Argentometric>=97.5 %
Melting point275°C to 280°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only