Thermo Scientific Chemicals

Dicumarol, 99%, Thermo Scientific Chemicals

Catalog number: 204120250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Dicumarol, 99%, Thermo Scientific Chemicals

Catalog number: 204120250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 204120250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
66-76-2
IUPAC Name
4-hydroxy-3-[(4-hydroxy-2-oxo-2H-chromen-3-yl)methyl]-2H-chromen-2-one
Molecular Formula
C19H12O6
InChI Key
DOBMPNYZJYQDGZ-UHFFFAOYSA-N
SMILES
OC1=C(CC2=C(O)C3=CC=CC=C3OC2=O)C(=O)OC2=CC=CC=C12
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with TBAH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text