Thermo Scientific Chemicals

Betaine, 98%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 204241000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Betaine, 98%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 204241000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 204241000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
107-43-7
IUPAC Name
2-(trimethylazaniumyl)acetate
Molecular Formula
C5H11NO2
InChI Key
KWIUHFFTVRNATP-UHFFFAOYSA-N
SMILES
C[N+](C)(C)CC([O-])=O
Infrared spectrum
Conforms
Iron (Fe)
=<20 ppm
Appearance (Color)
White
Titration with HClO4
97.5 to 102.5 %
Heavy metals (as Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text