Thermo Scientific Chemicals

5-Bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 204300250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Bromopyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 204300250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 204300250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4595-59-9
IUPAC Name
5-bromopyrimidine
Molecular Formula
C4H3BrN2
InChI Key
GYCPLYCTMDTEPU-UHFFFAOYSA-N
SMILES
BrC1=CN=CN=C1
Melting point
69°C to 73°C
Appearance (Color)
Yellow to beige to light brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text