Thermo Scientific Chemicals

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 204340250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 204340250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 204340250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3575-32-4
IUPAC Name
N1,N1-dimethylbenzene-1,3-diamine dihydrochloride
Molecular Formula
C8H14Cl2N2
InChI Key
BZJPIQKDEGXVFG-UHFFFAOYSA-N
SMILES
Cl.Cl.CN(C)C1=CC=CC(N)=C1
Appearance (Color)
White to beige to grey to purple
Titration Argentometric
>=98.5 %
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text