Solubility(5 % in water) Clear colorless to yellow to purple-brown
Appearance (Form)Powder
Titration with HClO498.5 to 101.5 %
Appearance (Color)White to yellow to beige to pink to grey
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.