Eicosane, 99%, Thermo Scientific Chemicals
Eicosane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Eicosane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 204370020
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.10 
Save 3.45 (13%)
-
Add to cart
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.10 
Save 3.45 (13%)
Add to cart
Eicosane, 99%, Thermo Scientific Chemicals
Catalog number204370020
Price (USD)/ Each
23.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
27.10 
Save 3.45 (13%)
-
Add to cart
Chemical Identifiers
IUPAC Nameicosane
Molecular FormulaC20H42
InChI KeyCBFCDTFDPHXCNY-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol)282.56
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Melting point36°C to 38°C
Infrared spectrumConforms
Appearance (Color)Transparent white
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only