1-Fluoro-3-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals
1-Fluoro-3-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Fluoro-3-iodobenzene, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 204480250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1121-86-4
IUPAC Name1-fluoro-3-iodobenzene
Molecular FormulaC6H4FI
InChI KeyVSKSBSORLCDRHS-UHFFFAOYSA-N
SMILESFC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
StabilizerCopper
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only