Thermo Scientific Chemicals

D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 204571000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 204571000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 204571000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14984-34-0
IUPAC Name
sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate
Molecular Formula
C6H9NaO7
InChI Key
WNFHGZLVUQBPMA-JSCKKFHOSA-M
SMILES
[Na+].O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C([O-])=O
Appearance (Form)
Crystalline powder
Water
6.7 to 8.7 % (K.F.)
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Specific optical rotation
+20° to +22° (20°C, 589 nm) (c=2, H2O)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text