N,N-Dimethylglycine hydrochloride, 99%
N,N-Dimethylglycine hydrochloride, 99%
Thermo Scientific Chemicals

N,N-Dimethylglycine hydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 204660050
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.30 
Save 6.65 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.30 
Save 6.65 (14%)
Add to cart
N,N-Dimethylglycine hydrochloride, 99%
Catalog number204660050
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.30 
Save 6.65 (14%)
-
Add to cart
Chemical Identifiers
CAS2491-06-7
IUPAC Namehydrogen 2-(dimethylamino)acetic acid chloride
Molecular FormulaC4H10ClNO2
InChI KeyFKASAVXZZLJTNX-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point188°C to 198°C
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only